VWR® Plastic Laminate Worksurface
Supplier: VWR International
Certificates
About this item
Laminate worksurfaces are composed of high-pressure plastic laminate.
- For High-Column worksurface two or three cut-outs accomodate 76" high columns, depending on bench width
- High-Column worksurfaces may be used with sitting/bench height carts when height-adjustment is required
Each worksurface has three square edges and one full 180-degree wrapped front edge.
Customers Who Bought This Also Bought