A propos de cet article
Glassware washers feature stainless steel construction for long life and corrosion protection.
- Up to four loading levels in two independently controlled chambers
- Each chamber can be customized to meet specific requirements
- Ultimate space saver provides up to 33% smaller footprint than washers of similar capacity
- Stainless steel construction ensures long life and corrosion protection
- Three separate filters protect pumps from debris and are conveniently located for easy access
- Delayed start feature allows washer to operate unattended
- Enhanced dual fan drying system
- Temperature selection to 70°C (158°F) for optimal energy savings of different loads
- Spindle Rack Ready models can accommodate 1 or 2 direct injection spindle racks
Multiple cycle selections and microprocessor control systems ensure that each load is processed efficiently with minimum energy consumption.
Featuring a high efficiency water management system to help conserve energy, these durable washers have a stacked double washing chamber with independent microprocessor controls for maximum throughput and flexibility. New improved units feature a soft touch controller, improved drying, a quieter run, and are compatible with existing racks and accessories. Three Vertical Stacked Washer models are available: Vertical Stacked Washer with Standard Rack for use with tap water, Vertical Stacked Washer with Top and Bottom DI Rinse - Spindle Rack Ready, and Vertical Stacked Washer with Top DI Rinse/Standard Rack and Bottom Spindle Rack Ready.
Spécifications
- Catalog No:
- 10002-866
- 10002-862
- 10002-864
- Electrical:
- 115 V, 60 Hz, 15 A
- 115 V, 60 Hz, 15 A
- 115 V, 60 Hz, 15 A
- Height:
- 182.88 cm (72.0")
- 182.88 cm (72.0")
- 182.88 cm (72.0")
- Depth:
- 55.0 cm (21.7")
- 55.0 cm (21.7")
- 55.0 cm (21.7")
- Width:
- 59.6 cm (23.5")
- 59.6 cm (23.5")
- 59.6 cm (23.5")