VWR® Undercounter Glassware Washers
Supplier: VWR International
Certificates
About this item
Economical glassware washers feature stainless steel construction for long life and corrosion protection.
- Stainless steel construction ensures long life and corrosion protection
- Insulated external casing
- Designed for optimum heat and noise absorption
- Three separate filters protect pumps from debris
- Multiple cycle selections and microprocessor control systems
- Enhanced dual fan drying system
- Temperature selection to 70 °C (158 °F) for optimal cleaning of different loads
- Delayed start feature
- Spindle rack ready models can accommodate 1 or 2 direct injection spindle racks
With adjustable legs, they can fit into any standard cabinet. New improved units feature a soft touch controller, improved drying, a quieter run, and are compatible with existing racks and accessories. Engineered for reliable and energy efficient operation, these durable washers are available in three models: Standard undercounter washer, undercounter washer with DI rinse, and undercounter washer with DI rinse/spindle rack ready.
Undercounter models are fully insulated for noise reduction but do not include side, top or rear casing.
Certifications: ETL and ETL-C listed.
Ordering information: A wide range of accessories are available to fit both upper and lower racks; spindle racks, specialty baskets, and other inserts. For more information, contact your Avantor representative.
Delivery: All models come with top and bottom stainless steel open racks to accommodate a variety of glassware; spindle racks purchased separately.
Caution: Notwithstanding any other Avantor terms and conditions of sale, an order of this product may not be cancelled and the product may not be returned except due to product defects or receipt of an incorrect product.
Specifications
- Catalog No:
- 10002-854
- 10002-850
- 10002-858
- Electrical:
- 115 V, 60 Hz, 15 A
- 115 V, 60 Hz, 15 A
- 115 V, 60 Hz, 15 A
- Height:
- 86.0 - 91.2 cm (33.9 - 35.9")
- 86.0 - 91.2 cm (33.9 - 35.9")
- 86.0 - 91.2 cm (33.9 - 35.9")
- Depth:
- 55.0 cm (21.7")
- 55.0 cm (21.7")
- 55.0 cm (21.7")
- Width:
- 59.6 cm (23.5")
- 59.6 cm (23.5")
- 59.6 cm (23.5")