Acid Cabinets, Poly or Wood Laminate, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
Durable and highly chemical-resistant.
- Safely store highly corrosive liquids such as phenol, nitric acid and sulfuric acid
- Ideal for smaller quantities at workbench level
- Haz-Alert™ reflective labels and double key set for safety and security
Long-lasting wood construction along with stainless steel handles and hinges provide exceptional resistance to corrosion from harsh acids.
Customers Who Bought This Also Bought