Staticide® Acrylic Conformal Coating, ACL
# de catalogue 75780-012
Fournisseur: ACL Staticide
Spécifications
- Volume:340 g (12 oz.)
- Description:Staticide® Acrylic Conformal Coating
- Cat. no.:75780-012
- Supplier No.:8690
Spécifications
A propos de cet article
Staticide® Acrylic Conformal Coating is ideal for providing insulation against high-voltage arcing and corona shorts
The Acrylic Conformal Coating fills holes quickly and easily. It is self healing, and contains more solids for a harder, longer lasting coat. The coating will not fragment, chip, or crack. It also provides exceptional acid resistance. Minimum dielectric strength is 1100 vol.ts/mil.
Operating temperature for this spray can is –59 to 132°C (–75 to 270°F). Contains UV tracer for easy quality control.
The coating provides a hard and durable protective barrier against humidity, salt, corrosive vapors, and fungal growth, which is essential for printed circuit boards and electronic assemblies as well as numerous other applications.
Certifications: Meets MIL-I-46058C, Type AR. Compliant to IPC-CC-830B Class B.
Customers Who Bought This Also Bought