ContactUsLinkComponent
Miriad Rapid HCV/HIV Antibody Test POU+, MedMira
Miriad Rapid HCV/HIV Antibody Test POU+, MedMira
Catalog # 75780-214
Supplier:  MEDMIRA
CAS Number:  
Miriad Rapid HCV/HIV Antibody Test 25tests
Miriad Rapid HCV/HIV Antibody Test POU+, MedMira
Catalog # 75780-214
Supplier:  MEDMIRA
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Miriad Rapid HCV/HIV Antibody Test POU+
  • Cat. no.:
    75780-214
  • Supplier No.:
    815311006012
Specifications
About this item
Miriad Rapid HCV/HIV Antibody Test employs MedMira Rapid Vertical Flow Technology to deliver the simultaneous diagnosis of Hepatitis C and HIV-1/2
Miriad HCV/HIV features a built-in reagent control line, works with whole blood, serum, and plasma specimens, and requires no additional equipment. Test kits can be stored for 12 months at 2–30°C (35.6–86°F).
In three easy steps users can obtain multiple instant results on a single test cartridge.
Ordering information: Each box of Miriad HCV/HIV POU+ Format includes 25 unitized test kits containing 1 test cartridge, 1 InstantGold cap, 1 Buffer Vial 1, 1 Buffer Vial 2, 1 transfer pipette and a package insert.
HBc/HIV/HCV Antibody Test controls are available as an accessory to this test..
Customers Who Bought This Also Bought