ISO 2431 Viscosity Cup, Industrial Physics
Fournisseur: GREAT LAKES GAGES, LLC
Certificats
A propos de cet article
The ISO 2431 Viscosity Cup is available in a range of titanium anodized aluminum or stainless steel formats
- A relatively deep well surrounding the top of the cup serves to catch any overflow
- The design of the cup and orifice eliminate hard to clean recesses
- The outside dimensions have been chosen to support the TQC stands
- Cups are manufactured under continuing quality control procedures
- Each cup features a unique, engraved serial number
These viscosity cups feature a fixed stainless steel nozzle (inner cavity). Laboratory type, to be used with stand, to measure the viscosity of lacquers, paint, and other liquids. Each immersion cup is supplied with a hard plastic storage case, with protective soft material on the interior.
Certifications: DIN, EN, ISO 2431.
Ordering information: Not calibrated. Calibration is available as an option; contact your Avantor representative for more information.
Customers Who Bought This Also Bought