DIN 53211 Immersion Cup, Industrial Physics
Fournisseur: GREAT LAKES GAGES, LLC
Certificats
A propos de cet article
The DIN 53211 Immersion Cups are available in a range of titanium anodized aluminum or stainless steel formats, all with a fixed stainless steel nozzle (inner cavity) and handle
- Each cup has a long handle, allowing it to be dipped by hand into a liquid container
- Design of the cup and orifice eliminate hard to clean recesses
- Cups are manufactured under continuing quality control procedures
- Each cup features a unique, engraved serial number
Inner dimensions are similar to DIN 53211. Cups are ideal for measuring coatings and other fluids during application or production. Each immersion cup is supplied with a hard plastic storage case, with protective soft material on the interior.
Certifications: Compatible with/similar to DIN 53211.
Customers Who Bought This Also Bought