Ford Viscosity Flow Cup, Industrial Physics
Fournisseur: GREAT LAKES GAGES, LLC
Certificats
A propos de cet article
Viscosity Flow Cups are used for measuring the consistency of paints, varnishes, and other similar products
- A relatively deep well surrounding the top of the cup serves to catch any overflow
- The design of the cup and orifice eliminate hard to clean recesses
- The outside dimensions have been chosen to support the TQC stands
- Cups are manufactured under continuing quality control procedures
- Each cup features a unique, engraved serial number
Cups are available in titanium anodized aluminum or stainless steel formats. All cups feature a fixed stainless steel nozzle (inner cavity). These viscosity cups are suitable for laboratory use with a stand.
Certifications: Compatible with ASTM D 1200.
Customers Who Bought This Also Bought