Ford Immersion Viscosity Cup, Industrial Physics
Fournisseur: Avantor
Certificats
A propos de cet article
The TQC Ford Immersion Cups are available in titanium anodized aluminum or stainless steel
- Each cup has a long handle, allowing it to be dipped by hand into a liquid container
- Design of the cup and orifice eliminate hard to clean recesses
- Cups are manufactured under continuing quality control procedures
- Each cup features a unique, engraved serial number
Both material formats of these viscosity cups feature a stainless steel inner cavity fixed nozzle and handle. Ideal for measuring coatings and other fluids during application or production. Each viscosity cup comes with a hard plastic storage case, with protective soft material on the inside.
Certifications: Compatible with ASTM D 1200.
Customers Who Bought This Also Bought