Zahn Immersion Viscosity Cup, Industrial Physics
Fournisseur: GREAT LAKES GAGES, LLC
Certificats
A propos de cet article
The TQC Zahn Immersion Cups are stainless steel viscosity cups with fixed stainless steel inner cavity nozzle and handle
- Each cup has a long handle, allowing it to be dipped by hand into a liquid container
- Design of the cup and orifice eliminate hard to clean recesses
- Cups are manufactured under continuing quality control procedures
- Each cup features a unique, engraved serial number
Ideal for measuring coatings and other fluids during application or production. Each viscosity cup is supplied with a hard plastic storage case, with protective material on the case interior.
Certifications: Complies with ASTM D1084 D4212.
Customers Who Bought This Also Bought