Biohazard Waste Cans, Justrite®
Supplier: Justrite
Certificates
About this item
Leakproof waste cans with self-closing lids provide safe, temporary collection of biohazardous waste.
- Great for medical facilities and research labs
- Foot-operated, self-closing lid reduces risk of contamination
- Reduce closure noise with SoundGard™ models
- Sturdy design with steel construction
Safely contain biohazardous materials, including bloodborne pathogens and other potentially infectious materials (no sharps).
Meets OSHA 29 CFR 1910.1030 for the segregation of infectious waste and FM approved for use with alcohols and other flammable liquids.
Certifications: FM, TUV, OSHA.
Ordering information: Includes OSHA biohazard label on front; use optional label kit 25880 for additional labels to add to the sides, to meet California Health and Safety Code.
Customers Who Bought This Also Bought