Type I Poly Safety Cans, Justrite®
# de catalogue 10836-908
Fournisseur: Justrite
Spécifications
- Capacity:1 gal.
- Description:Safety Can, 1G White Oval Poly
- Cat. no.:10836-908
- Supplier No.:12162
Spécifications
A propos de cet article
Chemical-resistance HDPE cans offer safe storage for flammable corrosives.
Certifications: NFPA, OSHA.
Customers Who Bought This Also Bought