Liquid Disposal Safety Cans, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
These high-density polyethylene (HDPE) disposal cans with built-in fill gauge provide superior chemical resistance and leak protection.
- Superior chemical resistance and leak protection
- Spring-loaded sealed cap automatically vents
- Stainless steel flame arrester
- Design resists accidental tip over
Built-in polyethylene fill gauge indicator pops up to warn you that the container is almost full.
Customers Who Bought This Also Bought