Spécifications
- Description:Pneumocystis jiroveci control slide
- Slide type:Fungi
- Cat. no.:10818-626
- Supplier No.:22251
Spécifications
A propos de cet article
Designed to be used with the Fungi Fluor Pneumocystis Kit
Slides are packaged 10 per box.
Unstained slides feature rat lung imprints and are methanol fixed. Store at room temperature. These control slides are compatible with other staining processes for P. Carinii, including Methenamine Silver Staining and Gram-Weigert techniques.