Air-Tight Damper, Labconco®
# de catalogue 82001-060
Fournisseur: Labconco
Product available on GSA Advantage®
Spécifications
- Description:Air-Tight Damper
- Height:25.4 cm (10")
- Cat. no.:82001-060
- Supplier No.:3776800
Spécifications
A propos de cet article
Mounts atop the Canopy Connection to adjust exhaust airflow and closes for decontamination procedures.
- Manually adjustable
- For use with Labconco Logic and Logic+ Biosafety Cabinets and Cell Logic and Cell Logic+ Biosafety Cabinets
Type 304 stainless steel. 10" (25.4 cm) high with 10" (25.4 cm) diameter OD inlet and outlet.
Customers Who Bought This Also Bought