Bottle, Wide Mouth, Glass, Raw, C&G Containers
Fournisseur: C&G Containers
Certificats
A propos de cet article
Glass bottles are not processed and they do not come with caps, either attached or in a separate pack.
- Bottles are not processed in any form
- Not pre-cleaned, assembled, nor certified
- They are 'raw' bottles
These bottles do not contain any type of documentation, and they do not contain a certificate of analysis in each product case.
The cases of these products also do not contain any box tags or labeling identifying them as C&G brand items. They do not feature a C&G part number or a custody seal. Only the manufacturer information will be stamped on each case of product.
Ordering information: Container measurements and capacity 'contain' volumes may vary.
Customers Who Bought This Also Bought