A propos de cet article
Heavy-gauge steel full-frame construction.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
- Designed to be used as a sink unit, but may be used anywhere wheelchair accessibility is needed
Cabinet with doors are constructed without a bottom, and with the toe-space rails attached to the doors so it will match adjacent cabinets when the doors are closed
Cabinet without doors are constructed without a bottom, and without doors for easy accessibility. The cabinet incorporates a 41⁄2" top rail to give a minimum of 271⁄2" clearance from the floor to the bottom of the rail and a removable sloping knee space panel to enclose piping and drain lines.
A 2.54 cm (1") thick work surface creates an overall height of 83.8 cm (33"). All cupboard cabinets are supplied with one full-width adjustable shelf.
The cabinet incorporates a 23⁄4" top rail to give a minimum of 291⁄4" of clearance from the floor to the bottom of the rail.