VWR® Contour™ Standing Height Pump Storage Cabinets
Fournisseur: VWR International
Certificats
A propos de cet article
Heavy-gauge steel full-frame construction.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- The interior is lined with 1" thick neoprene foam (for sound deadening)
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Each cabinet is furnished with a PVC vent pipe for venting to the fume hood above
A 2.54 cm (1”) thick work surface creates an overall height of 91.4 cm (36"). Cupboard cabinets are supplied with one full-width adjustable shelf.
(Requires a 2" hole in the hood work-surface). Each cabinet is furnished with 120 VAC, 20 amp, duplex receptacle mounted on the inside cabinet back and the pilot lighted toggle switch mounted in the top front rail (Wiring is not included).
Customers Who Bought This Also Bought