Pierce BupH™ Dry Blend Buffers, Washing, Thermo Scientific
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
BupH™ packs are pre-blended and pre-measured dry mixtures of buffers that are easy to prepare and used in protein purification and common laboratory techniques.
- Packs are pouches of dry-blend powder that are each sufficient to make 500 mL of buffer
- Eliminates weighing time and pH adjustments
- Every pack will yield the same, consistent buffer
- Eliminates concerns about long-term stability of stock solutions
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.