Shelf Storage Cabinets, Lista Xpress, Stanley
Fournisseur: Lista International
Certificats
A propos de cet article
Lista Xpress shelf cabinets, either with or without a lockable sliding door, are an efficient means for storing bulky items that don’t lend themselves to drawer storage.
- Open unit has 1 adjustable and 1 bottom shelf with no base
- Closable units have 2 lockable sliding doors and fork truck base
Open shelf cabinet’s interior and shelves match housing color. Sliding door cabinet’s interior matches housing color, and shelves are painted gray. Lockable unit cabinets have the choice of individual, keyed-alike or master key locks or no lock.
Customers Who Bought This Also Bought