ContactUsLinkComponent
Blueing reagent working solution, Pureview PH-Blue (pH 8)
Blueing reagent working solution, Pureview PH-Blue (pH 8)
# de catalogue 10809-908
Numéro CAS:  
Working Solution
Blueing reagent working solution, Pureview PH-Blue (pH 8)
# de catalogue 10809-908
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Blueing reagent working solution, Pureview PH-Blue (pH 8)
  • Size:
    500 ml
  • Cat. no.:
    10809-908
  • Supplier No.:
    16702-4
Spécifications
A propos de cet article
The Endure Staining System is designed to produce precise, consistent H&E staining.
A bluing reagent buffered to a constant pH 8. Fluctuations in pH have been associated with “floaters” and unclear nuclear detail. PH Blue is designed to be unaffected by carryover and is formulated to help standardize the staining process. Available in working solution or concentrate formula.
Customers Who Bought This Also Bought