Tissue Dye, Brush Tip, Cancer Diagnostics
Fournisseur: CANCER DIAGNOSTICS, INC.
Certificats
A propos de cet article
Brush tip tissue dye are designed for small biopsy use.
- Brush tip applicator
- Precise application
- Used for small biopsies
Brush tip applicator designed for small biopsies where precise application is required
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.