Spécifications
- Dimensions:26×20×1.5"
- Description:ESD Stacking Box, black
- Cat. no.:89022-942
- Supplier No.:8480005167
Spécifications
A propos de cet article
MFG Tray ESD stacking boxes provide optimal product protection with exceptional weight-carrying capability
Boxes possess superior physical properties over thermoplastic and corrugated products. They will not bend or sag under heavy loads and can be readily integrated into automated applications. ESD stacking boxes ensure optimum space utilization during assembly operations, and may be used to form mobile work stations. Fiberstat stacking boxes ensure complete protection from electrostatic discharge for the life of the product. They are resistant to cutting oils, greases, solvents, mild acids, and alkaline solutions within a pH rating of 3.0 to 10.0. Stacking boxes are easily cleaned in hot water or steam with standard detergents. Lids are available separately for certain models.