Spécifications
- Color:Gray
- Description:Dolly for MFG 870008, 875008, and 880008 series Stacking Containers
- Cat. no.:89022-912
- Supplier No.:8701485136
Spécifications
A propos de cet article
This heavy-duty fiberglass dolly will easily transport the content of MFG Tray's durable Stacking containers
These dollies and containers are resistant to cutting oils, grease, and solvents. Easy to clean with hot water or steam using standard detergents. Ideal for forgings, machined parts, stampings, ceramics, rubbers, and other heavy or hot parts emerging from molds or processing machines. Perfect for conveying heavy stacks of work-in-process parts on the standard 3” casters. MFG Tray is a leading manufacturer of high-strength, glass-reinforced composite containers, trays, boxes, flats, and totes for material handling in the metalworking, plastics, and electronics assembly industries, as well as in food service, confectionery, and pharmaceutical processing. MFG Tray products combine outstanding performance features with long service life to ensure exceptional value. The material matters in material handling.