ESD Shallow Textured Bottom Tray, MFG Tray
# de catalogue 89022-940
Fournisseur: Molded Fiber Glass Tray
Spécifications
- Dimensions:19.5 x 19.5 x 1.25"
- Color:Black
- Description:ESD Shallow Textured Bottom Tray
- pH range:3.0 to 10.0
- Temperature range:–51 to 121°C (–60 to 250°F), up to 300°F intermittently
- Cat. no.:89022-940
- Supplier No.:6290005167
Spécifications
A propos de cet article
Used in the assembly of circuit boards, computers, medical device, and other types of electronic or static sensitive assembly
Trays are designed for component assembly and for compatability with conveyors or cart transport systems. These trays feature a textured bottom for improved contact with conveyors.
Customers Who Bought This Also Bought