Mobile Storage Cabinets, Lista Xpress, Arlink
Fournisseur: Lista International
Certificats
A propos de cet article
Lista Xpress cabinets are durable, secure and designed to provide safe storage and transport for parts and tools
- Full extension drawers provides easy access and full use of the furthest corners of storage
- Individual drawer latches keep drawers closed while moving
- PrevenTip® locking system prevents accidental tipping
These cabinets includes mobile base with 2 fixed and 2 swivel 6 x 2" casters with wheel locks. These cabinets are lockable and come with individual lock and 2 keys or your choice of keyed-alike and master key systems.
Customers Who Bought This Also Bought