VWR® Contour™ Suspended Cabinets
Fournisseur: VWR International
Certificats
A propos de cet article
Heavy-gauge steel full-frame construction.
- Smooth, easy-to-maintain surfaces
- Phosphate-coated with a baked, chemical-resistant, powder-coat finish
- Manufactured of prime grade, cold-rolled, U.S. Standard sheet steel with no ragged edges
Cabinets are designed to hang beneath table frames. Light neutral color.
Customers Who Bought This Also Bought