Drawer Storage Cabinets, Lista Xpress, Arlink
Fournisseur: Lista International
Certificats
A propos de cet article
Lista Xpress cabinets are durable, secure and designed to provide years of safe, convenient storage and problem-free operation
- Fork truck base with front and rear covers
- Full extension drawers provides easy access and full use of the furthest corners of storage
- PrevenTip® locking system prevents accidental tipping
- Ergonomic handles improve safety and appearance
These drawers provides 440 lb. weight bearing capacity. Its divider features with ability to labelling, for the identification of compartment contents. These cabinets are lockable and come with individual lock and 2 keys or your choice of keyed-alike and master key systems.
Customers Who Bought This Also Bought