VWR® C-Leg Bench Frame with Top for Single Bay Uprights
Catalog # 10682-584
Supplier: VWR International
Specifications
- Worksurface Material:Laminate
- Cat. no.:10682-584
- Worksurface Thickness:1³⁄₁₆"
- W×D:72×32⅝"
Specifications
About this item
These VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
- Ergonomic and highly adaptable to the needs of the user
- Flexible, modular design
- Suitable for heavy loads (static) up to 1100 lbs.
- Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. All bench frames can be equipped with casters to create a mobile workstation solution. Frame color is light neutral.
Ordering information: VWR® lab benches are height adjustable in manual (without hand crank), hand crank, and motorized versions. Configurations include frame (pair of legs, rear case, and front rail) and top for single bay uprights (uprights not included). Top includes black phenolic or black laminate options.
Customers Who Bought This Also Bought