Spécifications
- Description:Undercounter freezer
- Environmentally Preferable:Safer Human and Environmental Health,Energy Efficient
- Model:FFS-04
- Cat. No.:10820-434
- Electrical:115 V, 60 Hz, 1.4A
- Shipping weight:55 kg (121 lb.)
- Defrost:Manual
- Door type:1 solid swing door
- Shelves/drawers:2 fixed shelves
- Exterior W×D×H:600×616×818 mm (23.625×24.25×32.188")
- Capacity:113 L (4 cu.ft.)
Spécifications
A propos de cet article
The VWR® Series Flammable Freezers are designed to meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations.
- Temperature controller
- Digital setting accuracy
- Precise temperature maintenance
- Independent operation for security
- Monitoring and alarms
- Meet NFPA guidelines
No electrical components are located in the interior of the unit, and the electrical components of the compressor have sealed in a vapor-proof enclosure for additional safety.
The 113 L (4 cu. ft.) unit uses Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
Units feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.