VWR® Filling and Venting Cap for VWR® Carboys
# de catalogue 10820-438
Fournisseur: VWR International
Spécifications
- Cap Size:83 mm
- Description:Filling/Venting 3-Port Cap for VWR® Carboys
- Material:PP/silicone
- Cap Color:Purple
Spécifications
A propos de cet article
The Filling and Venting Cap for VWR® Carboys is an autoclavable closure designed for the aseptic transfer of media, biological reagents, pure water, and chemicals
Each cap is equipped with integrated 1/4" ports.
Customers Who Bought This Also Bought