Frazier Suction Tube, OR Grade, Sklar®
Fournisseur: Sklar
Certificats
A propos de cet article
Sklar's® Frazier Suction Tube is a thin instrument used for removing fluid and debris from confined surgical sites such as the nasal cavity, lumbar and cervical regions or intracranially.
- Suction is controlled by a small hole on the handle
- Multiple tip sizes are available
- The size of the suction tube is indicated by the French measurement scale
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 10199-312
- 10199-314
- 10199-316
- 10199-318
- 10199-320
- 10199-304
- 10199-326
- 10199-306
- 10199-328
- 10199-330
- 10199-310
- 10192-464
- 10192-352
- 10192-374
- 10192-340
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free