Spécifications
- Length:6.9 cm (23/4")
- Description:Plain Wire Speculum Adult
- Material:Stainless Steel
- Curvature:Angled
- Cat. no.:10194-648
- Supplier No.:65-1127
Spécifications
A propos de cet article
Sklar's® Plain Wire Eye Speculum is an opthalmic instrument used to separate and hold open the eyelids
The tension in this spring wire device allows the lightweight, open wire to gently retracts both the top and bottom eyelids without excessive pressure on the globe. The choice of eye speculum is typically dependent upon the anatomical nature of the patient's eyes. This product is an adult speculum with 15×7 mm open blades.