Spécifications
- Sterility:Non sterile
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Material:Stainless Steel with Reduced Reflectivity Polymer Coating
- Size:Medium with Loop
- Tip Dimensions:4×11/4"
- Curvature:Straight
- Supplier No.:90-9055
Spécifications
A propos de cet article
The Sklar Black™ Graves Wide-View Vaginal Speculum is used for laser gynecological procedures
- Industry standard
It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Sklar Black™ Graves Vaginal Speculum is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.