PYREX® Nesbitt Absorption Bulb, Corning
Catalog # 89091-626
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Description:PYREX® Nesbitt Absorption Bulb
- Cat. no.:89091-626
- Supplier No.:1900-165
Specifications
About this item
Bulbs, made of PYREX® borosilicate glass are used for the absorption of CO₂ in carbon determination.
- Weighs approximately 135 grams when filled
- Reusable bulbs
- A turn of the interchangeable stopper closes both inlet and outlet tubes (Ext.Ø 6 mm)
Customers Who Bought This Also Bought