VWR® Disposable Sterile Boot Covers, SAL 10⁻⁶
Fournisseur: VWR International
Certificats
A propos de cet article
Disposable sterile boot covers are manufactured from 60gsm SMS fabric, and feature a 250gsm nonskid PVC sole.
- Disposable
- Sterile
- Vacuum packed in pairs
All boot covers feature bonded seams and elastic uppers for a secure fit. These garments are sterilized to a SAL of 10¯⁶. Boot covers are vacuum packed in pairs.
Certifications: ISO9001, ISO13485, ISO14001, CE, GMP.
Spécifications
- Catalog No:
- 10858-534
- 10858-532
- 10858-530
- Fabric:
- SMS/PVC
- SMS/PVC
- SMS/PVC
- Garment application:
- Cleanroom, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Cleanroom, Electronics/semi-conductor, Laboratory, Pharmaceutical
- Cleanroom, Electronics/semi-conductor, Laboratory, Pharmaceutical