PYREX® 105° Angled Tube Adapter with Two-Way 24/40 Standard Taper Joints , Corning
# de catalogue 89091-090
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- ST Joint:24/40
- Description:PYREX® 105° Angled Tube Adapter with Two-Way [ST] Joints
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:89091-090
- Supplier No.:8940-24
Spécifications
A propos de cet article
These angled two-way adapters are made of PYREX® borosilicate glass.
- Arms are approximately 105° apart
- Used as a receiver adapter
- Very good chemical and temperature resistance
One end is equipped with an outer 24/40 Standard Taper joint and the other end has an inner 24/40 Standard Taper joint. Used as a receiver adapter.
Customers Who Bought This Also Bought