PYREX® Reusable Volumetric Pipettes, Class A, Certified/Serialized, Corning
Supplier: Corning
Certificates
About this item
These reusable PYREX® glass pipettes are colour-coded by size for easy identification and sorting.
- Calibrated 'to deliver' (TD) their total capacity without blow-out
- Top end of 5 to 25 ml sizes is constricted (Mohr type)
- Coloured graduations are enameled onto the glass
- Colour-coded by size for easy identification
- Each pipette is individually serialised and certified
PYREX® Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Certifications: Calibrated to Class A tolerances in accordance with ASTM E-542, ATSM E-694, and ASTM E-1293 specifications.
Delivery: Supplied with a certificate of identification and capacity, traceable to NIST standards.
Specifications
- Catalog No:
- 53044-730
- 53044-738
- 53044-736
- 53044-734
- 53044-732
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey