PYREX® Bomb for Gum Stability, Corning
# de catalogue 52933-193
Fournisseur: Corning
Spécifications
- Description:Gum bomb (O.Ø×H = 50×105 mm)
- Cat. no.:52933-193
- Supplier No.:7732
Spécifications
A propos de cet article
Bomb with cover made from PYREX® borosilicate glass to determine gum stability in gasoline.
- Wide mouth with pourout style top
- Cover to prevent condensed vapours from contaminating the sample
- No interference with the free flow of oxygen
Replacement covers are available seperately.
Certifications: ASTM D-525
Customers Who Bought This Also Bought