PYREX® Claisen Three-Way Connecting Adapter, with 19/22 [ST] Joints, Corning
# de catalogue 32645-367
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Description:PYREX® Claisen Three-Way Connecting Adapter, with [ST] Joints
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Cat. no.:32645-367
- ST Joint Size:19/22
- Supplier No.:9050-19
Spécifications
A propos de cet article
These PYREX® Claisen three-way adapters have standard taper outer joints at the upper end and on the parallel sidearm, while an inner standard taper joint is positioned at the lower end.
- Top and sidearm [ST] outer joints
- Bottom [ST] inner joint
Adapters feature 19/22 standard saper outer joints at the upper end and on the parallel sidearm. An inner 19/22 standard taper joint is positioned at the lower end.
Customers Who Bought This Also Bought