PYREX® Reusable Serological Pipettes, for Cotton Plugging, TD, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Glass, reusable. These serological pipetes are designed for cotton plugging. The uniform top-end openings permit performing of plugs and allow precise control of liquid column.
- Color coded by size for easy identification and sorting
- Colored graduations are enameled onto the glass
- Calibrated to deliver (TD)
Graduation interval: 0.1 ml
Certifications: ASTM E-1044
Spécifications
- Catalog No:
- 77777-136
- 77777-138
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey