ContactUsLinkComponent
PYREX® Gas Washing Bottles with Coarse Fritted Disc, Corning
PYREX® Gas Washing Bottles with Coarse Fritted Disc, Corning
Supplier:  Corning
   Sustainable Options Available
Certificates
PYREX® Gas Washing Bottles with Coarse Fritted Disc, Corning
PYREX® Gas Washing Bottles with Coarse Fritted Disc, Corning
Supplier:  Corning
   Sustainable Options Available
Certificates
Specifications
  • Catalog No:
  • 16405-089
  • 16405-067
  • 16405-125
  • 16405-045
  • 16405-023
  • 16405-131
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Coarse frit pore distribution:
  • 40–60 µm
  • 40–60 µm
  • 40–60 µm
  • Coarse fritted disc diameter:
  • 60 mm
  • 60 mm
  • 60 mm
  • Extra coarse frit pore distribution:
  • 170–220 µm
  • 170–220 µm
  • 170–220 µm
  • Extra coarse fritted cylinder disc diameter:
  • 12 mm
  • 12 mm
  • 12 mm
  • Extra coarse fritted disc diameter:
  • 20 mm
  • 20 mm
  • 20 mm
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey