VWR® Trypsin (0.25%) EDTA (1X)
# de catalogue VWRL0154-0100
Fournisseur: VWR
Product available on GSA Advantage®
Numéro CAS:
Spécifications
- Size:100 ml
- Enzyme name:Trypsin
- Concentration:0.25%, 1X
- Formulation:solution in EDTA
- Cat. no.:VWRL0154-0100
Spécifications
A propos de cet article
Trypsin (0.25%) EDTA (1X) is used to release adherent cells from tissue culture plasticware for harvesting and passaging.
- Used to release adherent cells
- Aids in harvesting and passaging
Contains 0.001% phenol red, 0.25% Trypsin, 0.04% EDTA.
Customers Who Bought This Also Bought