PYREX® Dropping Bottle with Bulb and Pipette, Corning
# de catalogue 16359-060
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Capacity:125 ml
- Description:PYREX® Dropping Bottle with Bulb and Pipet
- Material:PYREX®
- Cat. no.:16359-060
- Supplier No.:1340-125
Spécifications
A propos de cet article
Narrow mouth droppings bottles, made of PYREX® borosilicate glass.
- Reusable bottle
- Pipette with suction bulb
- Hollow glass stopper style
Recommended for use where preventing contamination by solubility or pH change is important.
Customers Who Bought This Also Bought