PYREX® Weighing Bottle with External [ST] Style Opening, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Made of PYREX® borosilicate glass.
- Exterior joint improves material transfers and reduces risk of material contamination of cap
- Very good chemical resistance
- Very good temperature resistance
These standard weighing bottles have an external Standard Taper joint style opening. Bottle stoppers are hollow glass cap style over external ground body.
Spécifications
- Catalog No:
- 77776-952
- 77776-958
- 77776-960
- 77776-954
- 77776-956
- 77776-962
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey