Corning® Reusable Phenolic Caps, Corning
Fournisseur: Corning
Certificats
A propos de cet article
These reusable Corning® caps are fabricated from specially formulated phenolic resin to resist the effects of temperature and steam in autoclaving.
Caps have GPI threads and are provided with a glued-in white rubber liner.
Spécifications
- Catalog No:
- 60827-250
- 60827-271
- 60827-294
- 60827-237
- 60827-318
- 60827-330
- 60827-351
- 60827-409
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey