To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:tubulin alpha 1a, tubulin alpha 1b, tubulin alpha 1c
- Clonality:Polyclonal
- Gene ID:Q71U36/P68363/Q9BQE3/Q13748/Q6PEY2/P68366
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:TUBA1A, TUBA1B, TUBA1C
- Conjugation:Unconjugated
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:TUBA1A, TUBA1B, TUBA1C
- Modification:Tyr 272
- Storage buffer:Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Molecular weight:50-55 kDa
- Storage temperature:Store antibody at –20 °C for up to one year.
- Concentration:1 mg/mL
- Shipping temperature:4 °C
- Immunogen:TUBA1/3/4 (phospho Tyr272) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 272 (A-T-Y (p)-A-P) derived from Human TUBA1/3/4.
- Purification:Antibodies were purified by affinity-chromatography using epitope-specific peptide.
- Cat. no.:10814-080
- Supplier No.:79-938
Spécifications
A propos de cet article
Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulins. The genes encoding these microtubule constituents belong to the tubulin superfamily, which is composed of six distinct families. Genes from the alpha, beta and gamma tubulin families are found in all eukaryotes. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, which are highly conserved among species.
Western Blot: 1:500~1:1000
Type: Primary
Antigen: TUBA1A, TUBA1B, TUBA1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat