Buck Myringotome, OR Grade, Sklar
# de catalogue 10198-302
Fournisseur: Sklar
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Buck Myringotome
- Material:Stainless Steel
- Curvature:Straight
- Cat. no.:10198-302
- Supplier No.:67-2115
Spécifications
A propos de cet article
Sklar's Buck Myringotome, also known as a myringotomy knife, is used for opening the tympanic membrane to drain fluid and myringotomy procedures
This product is straight with a spearhead tip.
Customers Who Bought This Also Bought