Spécifications
- Sterility:Non sterile
- Description:Collin Vaginal Speculum
- Disposable / reusable:Reusable
- Latex/Latex-free:Latex-free
- Material:Stainless Steel
- Tip Configuration:41/4×11/4"
- Curvature:Straight
- Supplier No.:90-3945
Spécifications
A propos de cet article
Sklar's® Collin Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix.
- Double blade duckbill speculum inserted with the blades closed
- Industry standard
Unlike many other vaginal specula, there is no handle with the Collin device and the blades open laterally/horizontally versus vertically.